Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCGF6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PCGF6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15537220
![]() |
Novus Biologicals
NBP15537220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155372
![]() |
Novus Biologicals
NBP155372 |
100 μL |
Each for $487.50
|
|
|||||
Description
PCGF6 Polyclonal specifically detects PCGF6 in Human samples. It is validated for Western Blot.Specifications
PCGF6 | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
Q9BYE7 | |
84108 | |
Synthetic peptides corresponding to the middle region of human PCGF6 (polycomb group ring finger 6)(NP_115530). Peptide sequence: TQPLYNISANEGTGHFKPLEKKFVRVSGEATIGHVEKFLRRKMGLDPACQ. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
MBLRMel18 and Bmi1-like RING finger protein, MGC15678, MGC17541, polycomb group ring finger 6, RING finger protein 134polycomb group RING finger protein 6, RNF134Mel18 and Bmi1-like RING finger | |
PCGF6 | |
IgG | |
30 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title