Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PD 81723

Click to view available options
Quantity:
10 mg
50 mg
Description
Allosteric potentiator at the adenosine A1 receptor; acts via agonist-dependent and -independent mechanisms. Enhances agonist affinity for, and increased t½ of dissociation from, the receptor. Also inhibits basal and forskolin-stimulated adenylyl cyclase (AC) activity in A1 receptors expressed in CHO cells, possibly via direct potentiation of constitutive receptor activity or by direct inhibition of AC. Active in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 299.31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 299.311 |
| Chemical Name or Material | PD 81723 |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction