Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDZRN4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PDZRN4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP154925
![]() |
Novus Biologicals
NBP154925 |
100 μL |
Each for $487.50
|
|
|||||
NBP15492520
![]() |
Novus Biologicals
NBP15492520UL |
20 μL | N/A | N/A | N/A | ||||
Description
PDZRN4 Polyclonal specifically detects PDZRN4 in Human samples. It is validated for Western Blot.Specifications
PDZRN4 | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
DKFZp434B0417, IMAGE5767589, Ligand of Numb protein X 4, LNX4FLJ33777, PDZ domain containing ring finger 4, PDZ domain-containing RING finger protein 4, SAMCAP3L, SEMACAP3-like protein, SEMCAP3L | |
PDZRN4 | |
IgG | |
89 kDa |
Western Blot | |
Unconjugated | |
RUO | |
B4DG65 | |
29951 | |
Synthetic peptides corresponding to PDZRN4(PDZ domain containing ring finger 4) The peptide sequence was selected from the N terminal of PDZRN4. Peptide sequence SDSCHSLHPMEHEFYEDNEYISSLPADADRTEDFEYEEVELCRVSSQEKL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title