Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
| Assay Percent Range | 95% |
| Quantity | 250 mg |
| Formula Weight | 126.11 |
| Percent Purity | 97% |
| Chemical Name or Material | Perhydrocyclobuta[c]furan-1, 3-dione |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction