Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PGAM1 Antibody (1G4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB160864 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160864 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160864 Supplier Novus Biologicals Supplier No. NBP259418AF594
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PGAM1 Monoclonal antibody specifically detects PGAM1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen PGAM1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1G4
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B
Host Species Mouse
Immunogen Recombinant human PGAM1 (1-254aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5223
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.