Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PHT 427
Dual Akt and PDK1 inhibitor; antitumor
Specifications
Purity | >98% |
---|---|
Product Type | PHT 427 |
Molecular Weight (g/mol) | 409.61 |
Inhibitors | PHT 427 |
Description
Dual Akt and PDK1 inhibitor; binds the pleckstrin homology (PH) domain of Akt and PDK1 (Ki values are 2.7 and 5.2 μM, respectively). Inhibits Akt phosphorylation and Akt downstream targets; also induces apoptosis in vitro. Displays synergistic antitumor effect when combined with chemotherapeutics in breast cancer mouse models.Specifications
>98% | |
PHT 427 | |
409.61 | |
PHT 427 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title