Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Phyllite Specimen
SCI_ED

Catalog No. S26637 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Phyllite Specimen for demonstrations of metamorphic properties, such as glossy sheen and dense, foliated minerals.

  • Example of low-grade metamorphism from easily recognizable parent rock
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Fissile property of slate is retained, but has attained glossy sheen and soapy feel more indicative of metamorphic material

Specifications

Product Type Metamorphic Rock, Phyllite
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.3 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.