Learn More
Promega PI3K (p110alpha/p85alpha), 20ug, 200microl
Catalog No. prv1721 Shop All Promega Corporation Products
Click to view available options
Quantity:
200 μL
The PI3K lipid kinase enzymes are used with ADP-Glo KInase Assay System and Lipid Kinase Substrates in a homogeneous, add-mix-read lipid kinase assay.
  • Purified PI3K Class I Lipid Kinases
  • Use with ADP−Glo™ Assay and PI3K Substrates in lipid kinase activity assays
Specifications
Quantity 200 μL
Content And Storage <-65°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.