Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Picus™ NxT Electronic Pipette, 12 Channels, Trade-In

Catalog No. 14557832PR Shop All Sartorius Products
Click to view available options
Color:
Violet
Yellow
Volume (Metric):
5 to 120 μL
50 to 1200 μL

Upgrade your pipetting with unbeatable prices. For a limited time, you can get up to 35% savings on Sartorius Picus NxT Electronic Pipettes, 12 Channels.

Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.

  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management
  • Volume range: 0.2 μL to 1200 μL

Offer Details
After purchasing this trade-in program item please follow these steps to complete your trade in:
1. Complete Sartorius redemption form. (Contact Fisher Customer Service to obtain the form.)
2. Sartorius will email a link to a prepaid UPS shipping label, shipping instructions, and decontamination form.
3. Visit UPS site, print the shipping label, pack pipette, and ship to Sartorius.

Note: Any brand pipette, in any condition can be traded in. Pipettes must be free of contamination and returned together with the signed Decontamination Form.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Compatible Tips Universal Design, Safetyspace, Optifit
Autoclavable Not Autoclavable
Color Yellow
Disposable Reusable
For Use With ELISA, Protein Analysis and Cell Culture, PCR and Other DNA/RNA Techniques
Number of Channels 12
Product Line Picus™ NxT
Sterility Non-sterile
Type Electronic Pipettes
Volume (Metric) 5 to 120 μL
For Use With (Application) PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture
Warranty 2 Years
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.