Learn More
Sartorius Picus™ NxT Electronic Pipette, 12 Channels, Trade-In
Shop All Sartorius ProductsDescription
Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.
- Password protection for stored programs
- Very last liquid droplets dispensed by repeated blow-out for maximum recovery
- Features one-touch operating-button and feather-light electronic tip ejection
- Includes plate tracker for microwell plate dispensing
- Electronic piston control and brake for precise pipetting
- Safe-cone filters to prevent contamination
- Adjustment wheel for volume setting and navigation
- Range of pipetting modes for diluting and titrating to serial dispensing
- Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
- Certificate of accredited 3-point calibration
- RFID-enabled to support asset management
- Volume range: 0.2 μL to 1200 μL
Offer Details
After purchasing this trade-in program item please follow these steps to complete your trade in:
1. Complete Sartorius redemption form. (Contact Fisher Customer Service to obtain the form.)
2. Sartorius will email a link to a prepaid UPS shipping label, shipping instructions, and decontamination form.
3. Visit UPS site, print the shipping label, pack pipette, and ship to Sartorius.
Note: Any brand pipette, in any condition can be traded in. Pipettes must be free of contamination and returned together with the signed Decontamination Form.

Specifications
Specifications
Certifications/Compliance | CE Marked |
Compatible Tips | Universal Design, Safetyspace, Optifit |
Autoclavable | Not Autoclavable |
Color | Violet |
Disposable | Reusable |
For Use With | ELISA, Protein Analysis and Cell Culture, PCR and Other DNA/RNA Techniques |
Number of Channels | 12 |
Product Line | Picus™ NxT |
Sterility | Non-sterile |
Type | Electronic Pipettes |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.