Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molecular sieves 3A, 8 to 12 mesh
Click to view available options
Quantity:
500 g
5 kg
25 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
Specifications
Specifications
| Color | Beige to Brown |
| Quantity | 5 kg |
| Packaging | Plastic drum |
| Bulk Density | 600g/L min. |
| Water Adsorption | 20% min. (60% rel. humid.) |
| Residual Water | 1.5% max. |
| Physical Form | Beads |
| Solubility Information | Solubility in water: insoluble. |
| Chemical Name or Material | Molecular sieves 3A, 8 to 12 mesh |
Safety and Handling
| Hazard Category | HYGROSCOPIC |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction