Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLPP4/PPAPDC1A Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB341821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341821 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341821 Supplier Novus Biologicals Supplier No. NBP281931MFV610
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLPP4/PPAPDC1A Polyclonal antibody specifically detects PLPP4/PPAPDC1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PLPP4/PPAPDC1A
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Diacylglycerol Pyrophosphate Like 2, DPPL2, Phosphatidate Phosphatase PPAPDC1A, Phosphatidic Acid Phosphatase Type 2 Domain Containing 1A, Phospholipid Phosphatase 4, PPAPDC1
Host Species Rabbit
Immunogen PPAPDC1A antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PPAPDC1A. The immunogen is located within the last 50 amino acids of PPAPDC1A. Amino Acid Squence: ASLKKEERPTADSA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 196051
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.