Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pollen allergen Phl p 1 Antibody (Clone 25), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB342777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342777 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342777 Supplier Novus Biologicals Supplier No. NBP281276JF549
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Pollen allergen Phl p 1 Monoclonal antibody specifically detects Pollen allergen Phl p 1 in Plant samples. It is validated for Western Blot, ELISA, Dot Blot

Specifications

Antigen Pollen allergen Phl p 1
Applications Western Blot, ELISA, Dot Blot
Classification Monoclonal
Clone Clone 25
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Dot Blot
Formulation 50mM Sodium Borate
Gene Alias Allergen Phl p I, grass pollen allergen
Host Species Rabbit
Immunogen Recombinant Phl p 1, a grass pollen allergen.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.