Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ POROS™ HS 20 μm Strong Cation Exchange HPLC Columns
GREENER_CHOICE

Catalog No. 1332226
Click to view available options
Dimensions:
2.1 x 30 mm
4.6 x 100 mm
4.6 x 50 mm
Quantity:
1 Column

Catalog No. 13-322-26

Supplier: Thermo Scientific™ 1332226

Only null left
Add to Cart
Add to Cart

Thermo Scientific™ POROS™ Prepacked Strong Cation Exchange Column. POROS HS is a high capacity, strong cation exchange, High Performance Chromatography™ resins suitable for the separation and purification of biomolecules.

POROS HS is a high capacity, strong cation exchange, High Performance Chromatography™ resins suitable for the separation and purification of biomolecules. 20 micron particle size is used for high resolution and small scale preparative to semi-preparative separation of biomolecules.

POROS High Performance Chromatography™ Media and Pre-Packed Columns

  • A high performance chromatography resins for analytical to process scale separations.
  • Higher productivity: High throughput and high dynamic capacity associated with High Performance Chromatography™
  • Chemical stability: Allows aggressive cleaning and sanitization
  • Enhanced biomolecule access: Provided via large pores, ranging between 500-10,000 Å
  • Polystyrenedivinylbenzene particles: Yield a robust, easily packable matrix
  • Develop better separations methods in a shorter time frame: The speed of High Performance Chromatography technology reduces weeks of experimentation to only a few hours of work, so you have plenty of time to explore all the variables of your separation

Reduce time-consuming sample prep

You can replace many sample preparation steps with high speed High Performance Chromatography technology. For instance, dialysis of large volumes of material can be replaced by high flow rate processing of the dilute sample. Elution in a small volume of new buffer accomplishes both buffer exchange and concentration.

Create novel assays for more efficient analysis

High Performance Chromatography technology is not limited to standard modes of separation. Both enzymes and affinity ligands can be immobilized on POROS media. By combining rapid on-column protein digestions with rapid on-column immunoassays and chromatographic separations, you can create entirely new assays with unlimited potential.

High capacity, high resolution, high speed

In contrast to conventional chromatography media, POROS High Performance Chromatography™ resins particles, are engineered to have two discreet classes of pores. Large 'throughpores' allow convection flow to occur through the particles themselves, quickly carrying sample molecules to short 'diffusive' pores inside. By reducing the distance over which diffusion needs to occur, the time required for sample molecules to interact with interior binding sites is also reduced. Diffusion is no longer limiting and flow rates can be dramatically increased - without compromising resolution or capacity. Separations can be achieved at 1000 to 5000 cm/hr compared to 50 to 360 cm/hr for conventional media.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Particle Size 20 μm
Pore Size 2000 Å
Quantity 1 Column
Stationary Phase Sulfopropyl Polymer
Content And Storage Store at room temperature.
Product Line POROS
Binding Capacity 75 mg/mL (Lysozyme pH 6.2)
Shipping Condition Room Temperature
Volume (Metric) 1.7 mL
Dimensions 4.6 x 100 mm
Column Type Ion Exchange
Material PEEK
Type Prepacked Column
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.