Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.100 Normal (N/10) in Methanol, Reagents

Catalog No. 01336439
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Catalog No. Quantity
01-336-439 10 L
01-336-438 1 L
01-336-437 20 L
01-336-436 4 L
4 options

Catalog No. 01-336-439

Supplier: Reagents Holdings Llc C728505010F

Only null left
Add to Cart
Add to Cart

Potassium Hydroxide, 0.100 Normal (N/10) in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 1310-58-3 , 1310-58-3
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name methanol
SMILES CO

Specifications

pH 13
Color Colorless
Boiling Point 65°C
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 0.8
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.100 Normal (N/10) in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.