Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ PowerUp™ SYBR™ Green Master Mix for qPCR

Pre-formulated, optimized, universal 2X master mix for real-time PCR workflows.

Supplier:  Applied Biosystems™ A25777

Encompass_Preferred

 View more versions of this product

Catalog No. A25777


Get Up to $3,000 of qPCR Products with Purchase
View Available Promotions
Only null left
Add to Cart

Description

Description

PowerUp™ SYBR™ Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows. Coupled with user-supplied primer sets and template, PowerUp™ SYBR™ Green Master Mix is designed to amplifiy targets for accurate gene expression analysis.

PowerUp™ SYBR™ Green Master Mix features include:

  • A dual hot-start mechanism for excellent specificity
  • Highly reproducible CTs over a broad dynamic range
  • Inclusion of UDG to help prevent carryover contamination
  • Stability of pre-assembled reactions for up to 72 hours
  • Compatibility with most real-time qPCR instruments

Exceptional specificity with dual hot-start mechanism

Specificity is paramount in obtaining high-quality data in SYBR™ Green reactions and can be enhanced through control of the Taq DNA polymerase at lower temperatures before stringent primer binding. PowerUp™ SYBR™ Green Master Mix uses the Dual-Lock™ Taq DNA polymerase enzyme that combines two unique hot-start mechanisms to help control its activity and to prevent undesirable early activity of the polymerase at low temperatures. In an experiment evaluating 24 different primer sets, PowerUp™ SYBR™ Green Master Mix generated single melt curves 100% of the time, consistent with highly specific amplification.

Tight reproducibility in CTs over a wide dynamic range

PowerUp™ SYBR™ Green Master Mix demonstrates excellent reproducibility over a wide dynamic range for a variety of targets tested (see figure). In addition, this master mix provides efficient amplification over 6 logs of sample input. Unlike competitor mixes that can exhibit inhibition at high cDNA inputs and low correlation coefficients over a dynamic range experiment, PowerUp™ SYBR™ Green Master Mix efficiency is maintained over the full dynamic range to help deliver maximal confidence in data quality (see figure).

UDG included for carryover contamination control

The inclusion of uracil-DNA glycosylase (UDG) and dUTP in the PowerUp™ SYBR™ Green Master Mix enables the degradation of any previously amplified PCR products, helping prevent contamination of subsequent qPCR reactions and possible false positives.

Stability of pre-assembled reactions

The tight control of the Dual-Lock™ Taq DNA polymerase in PowerUp™ SYBR™ Green Master Mix enables reactions to be assembled up to 72 hours prior to cycling without impacting data (see figure). This consistency allows users to have confidence and flexibility in the use of PowerUP™ SYBR™ Green Master Mix across numerous workflow scenarios¹.

Broad instrument compatibility

PowerUp™ SYBR™ Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments². It is also compatible with the Bio-Rad IQ™5 and CFX96™ / CFX384™ systems, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems. For optimal results, recommended primer concentrations are 300–800 nM.

¹Pre-PCR stability is influenced not only by the master mix, but also by the target being analyzed. For maximum confidence, researchers should confirm stability profiles of their specific targets.

² For optimal performance, standard cycling using the Applied Biosystems™ 7900 system is recommended.

Order Info

Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2X
SYBR
qPCR
Fast or Standard
Single-plex
PowerUP™, SYBR™
5 x 5 mL
2500 Reactions of 20-μL
Gene Expression
2500 Reactions
5 x 5 mL
Medium
The 2X mix contains SYBR Green dye, Dual-Lock Taq DNA Polymerase, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye, and optimized buffer components. Contains 5 x 5 mL tube, sufficient for 2500 20-μL reactions (10 μL Master Mix per reaction).

Store at 2–8°C in the dark.
Tube
Dual-Lock Taq DNA Polymerase
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 7 Flex, QuantStudio™ Absolute Q Digital PCR System, StepOne™, StepOnePlus™, ViiA™ 7 System
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
Wet Ice
cDNA or gDNA only
72 hr., Assembled Reaction
DNA
dUTP, UNG/UDG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.