Learn More
Corepoint Scientific™ Premier Pharmacy Stainless Steel Laboratory Series Refrigerator
SDP
Catalog No. 22651154 Shop All Nor-Lake Scientific Products
Premier Pharmacy Stainless Steel Laboratory Series Refrigerator feature microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats.
  • Microprocessor temperature controller with audible and visual alarms
  • Digital temperature display
  • Freestanding thermometer with 3 years certificate of calibration
  • Two probes; 1 in air and 1 in sample bottle
  • Remote alarm contacts, keyed door locks, casters, and pharmacy refrigerator toolkit, temperature logs and CDC approved power cord labels
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.