Learn More
G-Biosciences Probe-Based One Step RT-qPCR 2X Master Mix
SCI_ED
Probe-based one step RT-qPCR 2X master mix is used for gene expression analysis, genotyping and mutation detection, pathogen detection, viral detection, absolute quantification. It has enhanced efficiency, specificity, sensitivity and compatible with nearly all real time PCR instruments.
Supplier:  G-Biosciences 7861987
 View more versions of this product
Catalog No. S03130
Only null left
Add to Cart
Description
Description
  • G-biosciences probe-based one step RT-qPCR 2X master mix contains Reverse Transcriptase, Taq DNA polymerase, low ROX reference dye, dNTPs and optimized buffer with stabilizers.
  • This master mix is ideal for high-throughput real-time PCR screening and validation which requires minimal handling and offer robust and consistent results.
Specifications
Specifications
Primer-probe
High
Fast or Standard
Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection
Liquid
1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, qPCR, RT-qPCR
RT-qPCR Master Mix
1000
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.