Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Probe-Based One Step RT-qPCR 2X Master Mix
SCI_ED

Probe-based one step RT-qPCR 2X master mix is used for gene expression analysis, genotyping and mutation detection, pathogen detection, viral detection, absolute quantification. It has enhanced efficiency, specificity, sensitivity and compatible with nearly all real time PCR instruments.

$200.00 - $3389.00

Specifications

Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, qPCR, RT-qPCR
Product Type RT-qPCR Master Mix
View More Specs

Products 4
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
S03129
SCI_ED
View Documents
G-Biosciences
7861988
5000
Each for $3,389.00
Only null left
Add to Cart
 
S03130
SCI_ED
View Documents
G-Biosciences
7861987
1000
Each for $753.00
Only null left
Add to Cart
 
S03131
SCI_ED
View Documents
G-Biosciences
7861986
500
Each for $430.00
Only null left
Add to Cart
 
S03132
SCI_ED
View Documents
G-Biosciences
7861985
200
Each for $200.00
Only null left
Add to Cart
 
Description

Description

  • G-biosciences probe-based one step RT-qPCR 2X master mix contains Reverse Transcriptase, Taq DNA polymerase, low ROX reference dye, dNTPs and optimized buffer with stabilizers.
  • This master mix is ideal for high-throughput real-time PCR screening and validation which requires minimal handling and offer robust and consistent results.
Specifications

Specifications

Primer-probe
High
RT-qPCR Master Mix
Liquid
1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, qPCR, RT-qPCR
Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.