Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-Biosciences Probe-Based One Step RT-qPCR 2X Master Mix

Click to view available options
No. of Reactions:
200
500
1000
5000
Description
- G-biosciences probe-based one step RT-qPCR 2X master mix contains Reverse Transcriptase, Taq DNA polymerase, low ROX reference dye, dNTPs and optimized buffer with stabilizers.
- This master mix is ideal for high-throughput real-time PCR screening and validation which requires minimal handling and offer robust and consistent results.
Specifications
Specifications
| Detection Method | Primer-probe |
| Form | Liquid |
| GC-Rich PCR Performance | High |
| PCR Method | 1-step RT-qPCR, 2-step RT-qPCR, Multiplex qPCR, qPCR, RT-qPCR |
| Reaction Speed | Fast or Standard |
| Product Type | RT-qPCR Master Mix |
| For Use With (Application) | Gene Expression, Genotyping, Microbial Detection, Pathogen Detection, Virus Detection |
| No. of Reactions | 500 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction