Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS Process NPD™ One-Step Germicidal Detergent
Process NPD™ one-step germicidal detergent is a quaternary ammonium disinfectant, a non-sterile, double bagged concentrate with no perfumes or dyes.
$632.50 - $3750.00
Specifications
Product Type | Germicidal Detergent |
---|---|
pH | 10.2 to 10.7 |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Volume (English) | Container Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Container Type | Price | Quantity & Availability | ||||
19-415-157
|
STERIS
639001 |
55 gal. | Drum |
Each for $3,750.00
|
|
||||
14-415-158
|
STERIS
639002 |
30 gal. | Drum |
Each for $2,680.00
|
|
||||
14-415-157
|
STERIS
639008 |
1 gal. | Plastic Bottle |
Case of 4 for $632.50
|
![]() |
||||
Description
- Non-sterile version of Process NPD™ family of one-step, quaternary ammonium cleaner/disinfectants made specifically for use on hard non-porous surfaces
- Made with same formulation that contains no perfumes or dyes that might interfere with critical research studies or manufacturing processes
- Use-dilution rate of 1:256
- Kill a broad spectrum of common bacteria without harming surface
How Process NPD™ One-Step Germicidal Detergent Works
- Liquid concentrate specifically formulated for disinfection of hard surfaces
- Use-dilution can be applied manually or sprayed
Why Process NPD™ One-Step Germicidal Detergent?
- Efficacy
- Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
- Productivity
- Formulation includes surfactants for surface cleaning while disinfecting
- Flexibility
- Ideal for use in contamination control rotation
- Reliability
- Complete lot traceability
- Assurance
- Each lot documented by certificate of analysis
Specifications
Germicidal Detergent | |
10.2 to 10.7 | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title