Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Procollagen I N-Peptide Monoclonal Antibody (C6)
Mouse Monoclonal Antibody
$243.00 - $588.50
Specifications
Antigen | Procollagen I N-Peptide |
---|---|
Clone | C6 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541662
|
Invitrogen™
MA541662 |
200 μL |
Each for $588.50
|
|
|||||
PIMA543883
|
Invitrogen™
MA543883 |
20 μL |
Each for $243.00
|
|
|||||
Description
Procollagen I N-Peptide Monoclonal Antibody for Western Blot, IHC (P)
Procollagen type I N-terminal propeptide (P1NP) is a specific marker of bone formation, reflecting the synthesis of type I collagen, which constitutes the major part of the bone matrix. P1NP is derived during the conversion of procollagen to collagen, produced by osteoblasts during bone formation. Its primary clinical application lies in the diagnosis and monitoring of metabolic bone diseases, particularly osteoporosis. In osteoporotic patients, P1NP levels can predict bone mineral density (BMD) responses to treatments like teriparatide and romosozumab. Low serum levels of P1NP are indicative of a high risk of BMD loss, providing a valuable tool for premenopausal women with systemic lupus erythematosus (SLE) and other conditions affecting bone health.Specifications
Procollagen I N-Peptide | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P02452 | |
1277 | |
Recombinant protein Procollagen I N-Terminal Propeptide. | |
Antibody |
C6 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
COL1A1 | |
P1NP; Procollagen I N | |
COL1A1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title