Learn More
Description
- Chemiluminescent Substrate to Detect Horseradish Peroxidase (HRP) Conjugates on Immunoblots
- Detect picogram levels of protein with minimal background
- Non-radioactive
- No optimization required; easily switch from other entry-level ECL substrates
Specifications
Specifications
| Product Type | ECL Western Blotting Substrate |
| Quantity | 250 mL |
| For Use With (Application) | Detects picogram amounts of Antigen and presence of HRP, blots can be repeteadly exposed to X-ray |
| Storage Requirements | 2°C to 10°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.