Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Exonuclease III

Catalog No. p-3509040 Shop All Promega Corporation Products
Click to view available options
Quantity:
5000 U

Includes

  • Part No. M1811: Exonuclease III 10X Buffer (1 x 1mL); Exonuclease III (5000U).
  • Part No. M1815: Exonuclease III 10X Buffer (5 x 1mL); Exonuclease III (25,000U).

Exonuclease III is a 3' → 5' exonuclease specific for dsDNA.

  • A 3′→5′ Exonuclease Specific for Double-Stranded DNA
  • Control deletion rate by varying incubation temperature
  • Inactivate Exonuclease III by heating at 75°C for 10 minutes
  • Supplied at a concentration of 150-200u/μL

Specifications

Concentration 150 to 200 U/μL
Color Colorless
Components 10X Reaction Buffer: 660mM Tris HCl (pH 8 at 25°C), 6.6mM MgCl2
Purity >90%
pH 7.5
For Use With (Application) Generating nested sets of deletions with double-stranded, linear DNA, preparation of single-stranded DNA
Storage Buffer 20mM Tris HCl (pH 7.5 at 25°C), 1mM DTT, 100mM KCl, 50% Glycerol
Content And Storage -30°C to -10°C
Source Recombinant E. coli strain
Quantity 5000 U
Product Type Exonuclease III
Molecular Weight (g/mol) 28000
Inhibitors 50mM NaCl
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.