Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HDAC-Glo™ I/II Assay and Screening Systems

Catalog No. p-4265688 Shop All Promega Corporation Products
Click to view available options
Quantity:
10 mL
50 mL
100 mL
Product Type:
Assay
Screening System

The HDAC-Glo™ I/II Assays and Screening System are single-reagent-addition, homogeneous, luminescent assays that measure the relative activity of histone deacetylase (HDAC) class I and II enzymes from cells, extracts or purified enzyme sources.

  • Detects HDAC Class I and II Enzyme Activity
  • Simple add-mix-measure protocol
  • Use purified enzymes, extracts or cells directly in culture plates
  • Screening System includes HeLa Nuclear Extract

Specifications

Quantity 100 mL
pH 8
Product Type Assay
Content And Storage -30°C to -10°C
For Use With (Application) Determining HDAC inhibitor potency using purified enzymes, extracts or cells directly in culture plates
Form Fluid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.