Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Isoquant™ Isoaspartate Detection Kit

Catalog No. prma1010 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
100 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRMA1010 100 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRMA1010 Supplier Promega Supplier No. MA1010
Only null left
Add to Cart
Add to Cart

Includes

1mL A117A Protein Isoaspartyl Methyltransferase, 300μL A233A S-Adenosyl-Homocysteine, 50μL A119A Isoasp-DSIP, Reference Standard, 1mL A232A Stop Solution NR, 0.5mL A120A S-Adenosyl-L-Methionine, 1mL A118A Reaction 5X Buffer

ISOQUANT Isoaspartate Detection Kit is intended for quantitative detection of isoaspartic acid residues in proteins and peptides.

  • Quantitative Detection of Isoaspartic Acid Residues in Proteins and Peptides
  • Monitor Protein Deamidation during Storage and Production
  • Environmental and Industrial Uses

Specifications

For Use With (Application) For analysis of isoaspartate residues in Peptides or Proteins
Content And Storage −30 to −10°C
Product Type Detection kit
Quantity 100 Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.