Learn More
Description
- Inhibits Activation of MAPK (ERK 1/2)
- Used in both in vivo and in vitro studies of MEK
- Potent: inhibits Raf-activated MEK1/2 in vitro with an IC50 of ∼10-20microM
- Supplied as a dried material
Specifications
Specifications
| For Use With (Application) | Used in both in vivo and in vitro studies of MEK |
| Content And Storage | −30 to −10°C |
| Product Type | MEK inhibito r U0126 |
| Quantity | 5 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.