Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Modifying Enzymes: Single-Stranded DNA Binding Protein

Catalog No. p-127798 Shop All Promega Corporation Products
Click to view available options
Quantity:
100 μg

Binds with high affinity in a cooperative manner to single-stranded DNA but does not bind well to double-stranded DNA

  • Cooperatively Binds ssDNA with High Affinity
  • Consists of four identical 18.9kDa subunits
  • Does not bind well to double-stranded DNA
  • Supplied at a concentration of 1-5μg/μL

Specifications

Concentration 1 to 5 μg/μL
Color Colorless
Components 4 Identical 18.9kDa subunits
pH 7.4
For Use With (Application) Studying DNA replication and recombination, DNA sequencing
Storage Buffer 50mM Tris HCl (pH 7.4), 1mM EDTA, 1mM DTT, 0.2M NaCl, 50% Glycerol
Content And Storage -30°C to -10°C
Source E. coli
Quantity 100 μg
Product Type Single-stranded DNA Binding Protein
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.