Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MULTI-CORE™ Buffer Pack

Catalog No. prr9991 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
Catalog No. Quantity
PR-R9991 3 x 1 mL
1 options

Catalog No. PR-R9991

Supplier: Promega R9991

Only null left
Add to Cart
Add to Cart

Universal restriction enzyme buffer. Supports double-digests with many restriction enzymes.

  • Convenient Aliquots of the Promega Universal Restriction Enzyme 10X Buffer
  • Enables digestion of DNA with more than one enzyme in a single reaction
  • Only modest adjustments in the amount of enzyme used will ensure complete multiple digestions

Specifications

Color Colorless
Components Universal restriction Enzyme 10X Buffer
pH 7.7
For Use With (Application) Performing multiple digestions of DNA with more than one Enzyme in a single reaction
Content And Storage -30°C to -10°C
Quantity 3 x 1 mL
Product Type Buffer Pack
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.