Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega P450-Glo™ CYP1A2 Induction/Inhibition Assay

Catalog No. p-4390042 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8421 10 mL
PRV8422 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8421 Supplier Promega Supplier No. V8421
Only null left
Add to Cart
Add to Cart

Includes

Luciferin-1A2 (30μL), 500X D-Cysteine (100μL), luciferin detection reagent and reconstitution buffer.

The P450-Glo™ CYP1A2 Assay and Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays

  • A Luminescent Assay and Screening System for Measuring CYP1A2 Activity
  • Screening system includes a complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background

Specifications

Quantity 10 mL
Content And Storage -30°C to -10°C
Detection Method Homogeneous, Luminescence
For Use With (Application) For measuring cytochrome P450 activity
Kit Contents 1 x 30μL Luciferin-1A2, 1 x 100μL D-Cysteine 500X, 1 x 1 each Luciferin detection reagent, 1 x 10mL Reconstitution Buffer
For Use With (Equipment) For 384-well format
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.