Learn More
Promega ProFluor™ SerThr Phosphatase Assay
Catalog No. p-2819229 Shop All Promega Corporation Products
Click to view available options
Quantity:
4 Plate
8 Plate
The ProFluor Ser/Thr PPase Assay measures purified serine/threonine protein phosphatase activity in a multiwell plate format and is ideal for high-throughput applications.
  • Fluorescent, “Add-Mix-Read” Ser/Thr Phosphatase Assay
  • Works with protein phosphatase 1 (PP1), PP2A, PP2B and PP2C
  • Ideal for high-throughput phosphatase assays in 96- or 384-well plate formats
  • Minimal test compound interference
Patent pending. This product is covered by U.S. Patents 4,557,862 and 4,640,893 and is sold for research use only. All other uses, including, but not limited to, use as a clinical diagnostic or therapeutic, require a separate license. Please contact Prom
Specifications
Description ProFluor Ser/Thr Assay System
Content And Storage −30 to −10°C
Quantity 8 Plate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.