Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Pvu I (D)

Catalog No. p-2387569 Shop All Promega Corporation Products
Click to view available options
Quantity:
100 U
500 U
Concentration:
2 to 10 U/μL
Cut Site:
CGAT.CG
CGAT.CG GC.TAGC

Catalog No. PR-R6325

Supplier: Promega R6325

Only null left
Add to Cart
Add to Cart

Active and capable of digestion directly in GoTaq™ Green Master Mix. (CG AT/CG GC\TA GC)

  • Active and capable of digestion directly in GoTaq(R) Green Master Mix

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 2 to 10 U/μL
Components 1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U PvuI
Incubator Temperature 37°C
pH 7.3
For Use With (Application) Active and capable of digestion directly in GoTaq™ Green Master Mix
Storage Buffer 10mM Tris HCl (pH 7.3 at 25°C), 50mM NaCl, 0.1% Triton™ X-100, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 500 U
Cut Site CGAT.CG
Product Type PvuI
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.