Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Ssp I Genome Qualified (E)

Catalog No. p-2387584 Shop All Promega Corporation Products
Click to view available options
Quantity:
2500 U
Concentration:
40 to 80 U/μL

Genome qualified to ensure optimal performance in genomic analysis applications. (AAT/ATT TTA\TAA)

  • Genome qualified to ensure optimal performance in genomic analysis applications

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 40 to 80 U/μL
Components 1 x 1mL 10X Buffer E, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U SspI (HC), 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Assayed to ensure optimal performance in genomic analysis applications
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -20°C
Quantity 2500 U
Cut Site AAT.ATT TTA.TAA
Product Type SspI (HC)
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.