Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzymes - Mbo II (B)
Shop All Promega Corporation Products
Click to view available options
Quantity:
100 U
Description
- Incubate buffer B at 37°C
- Moraxella bovis ATCC 10900
- 10mM Tris-HCl (ph 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/ml BSA, 50% glycerol
- Store at -20°C
Sensitive to overlapping dam methylation, MboII produces a single-base overhang, which is difficult to ligate
Specifications
Specifications
Concentration | 2 to 10 U/μL |
Components | 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U MboII, 1 x 0.25mL Muti-Core™ 10X Buffer |
Incubator Temperature | 37°C |
pH | 7.3 |
For Use With (Application) | Enzymes that recognize short, specific DNA sequences in which the Enzymes then cleave |
Storage Buffer | 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
Quantity | 100 U |
Cut Site | GAAGA(N)8. CTTCT(N)7 |
Product Type | MboII |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction