Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega x-tracta™ Gel Extractors

Catalog No. p-4532429 Shop All Promega Corporation Products
Click to view available options
Quantity:
100/Pk.
25/Pk.

The x-tracta Gel Extractor tool provides a convenient, safe method for removal of agarose gel fragments for further processing. The device removes a 0.13 x 0.33 inch gel piece from agarose gels for easy transfer into a microcentrifuge tube for processing.

  • Safe, Convenient Extraction of Gel Slices
  • Removes a 0.13 x 0.33 inch gel piece
  • Disposable device reduces cross-contamination
  • No need to use razor blades or scalpels

Specifications

Type x-tracta™ Gel Extractor
Quantity 25/Pk.
Temperature (Metric) Storage 15°C to 30°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.