Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROP1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7247031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166108 20 μg
NB166107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166108 Supplier Novus Biologicals Supplier No. NBP31550620UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PROP1 Polyclonal antibody specifically detects PROP1 in Mouse samples. It is validated for Western Blot

Specifications

Antigen PROP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CPHD2, homeobox protein prophet of Pit-1, Pituitary-specific homeodomain factor, PROP paired-like homeobox 1, PROP-1, prophet of Pit1, paired-like homeodomain transcription factor
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human PROP1 (NP_006252.3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 5626
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.