Learn More
Thermo Scientific™ ProPac™ PA1 HPLC Columns
Description
Efficiently analyze and purify hydrophilic anionic proteins and peptides with Thermo Scientific™ ProPac™ PA1 LC Columns. These strong anion-exchange columns provide high-resolution separations of proteins with pI values from 3 to 11.
The columns can also resolve some proteins and peptides that are highly cationic and have very high isoelectric points.
- Good for hydrophilic proteins and peptides
- Ideal for high-resolution separations of proteins with pI values from 3 to 11
- Pellicular packing ensures high-efficiency and fast mass transport

Specifications
Specifications
Column Format | Analytical Column |
Diameter (Metric) | 9 mm |
Flow Rate | 0.9 to 1.5 mL/min |
For Use With | BioLC Chromatography System |
Length (Metric) | 250 mm |
Max. Pressure | 5000 psi (345 bar) |
Particle Size | 10 μm |
Pore Size | Nonporous |
Quantity | Each |
Stationary Phase | Alkyl Quaternary Amine |
Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.