Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eBioscience™ Propidium Iodide Staining Solution
Click to view available options
Quantity:
2 mL
Description
- 5μL (40 ng)/test
- Can be used in FL3 for inviability exclusion, but should be analyzed in FL2 when used as a counterstain for FITC Annexin V
Specifications
Specifications
| Color | Red |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Staining Solution |
| Dye Type | Cell-Impermeant |
| Emission | 535/617 |
| Product Line | eBioscience |
| Quantity | 2 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction