Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein phosphatase 1 inhibitor 3D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Protein phosphatase 1 inhibitor 3D |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Protein phosphatase 1 inhibitor 3D Polyclonal specifically detects Protein phosphatase 1 inhibitor 3D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Protein phosphatase 1 inhibitor 3D | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
5509 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ELAQVKVFNAGDDPSVPLHVLSRLAINSDLCCSSQDLEFTLHCLVPDFPPPVEAADFGERLQRQLVCLERVTCSDL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
DKFZp781L2441, PP1 subunit R6, PPP1R6, protein phosphatase 1 regulatory subunit 3D, protein phosphatase 1 regulatory subunit 6, protein phosphatase 1, regulatory subunit 3D, protein phosphatase 1, regulatory subunit 6, spinophilin, protein phosphatase 1-binding subunit R6, regulatory (inhibitor) subunit 3D | |
PPP1R3D | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title