Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Proteinase K (PK) Solution

Catalog No. prmc5005 Shop All Promega Corporation Products
Click to view available options
Quantity:
4 mL
16 mL

Catalog No. PRMC5005

Supplier: Promega MC5005

Only null left
Add to Cart
Add to Cart

Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. Proteinase K (PK) Solution is supplied at a concentration of 20mg/ml in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride, 50% glycerol.

  • Stable at Room Temperature and Easy to Use
  • Active over pH range 4.3-12.0 in 0.5% SDS or 1% Triton(R) X-100
  • Retains >80% activity at temperatures up to 60°C
  • No resuspension or thawing before use
  • Cat.# MC5005, MC5008 supplied at a concentration of 20mg/ml

Specifications

Concentration 20 mg/mL
pH 4.3 to 12
Quantity 4 mL
Product Type Proteinase K (PK) Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.