Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ PRP™-X300 Ion Exclusion HPLC Columns

Catalog No. p-51835 Shop All Hamilton Company Products
Click to view available options
Length (Metric):
250 mm

Polymeric column packing for the separation of alcohols and organic acids

A fast, versatile HPLC column packing material, Hamilton PRP-X300 columns rapidly and easily separate closely related alcohols and organic acids without column heating. The sulfonated poly(styrene-divinylbenzene) support separates samples by a mixed mode mechanism.
Three Separation Modes

  • Hydrogen Bonding, Reversed Phase, and Ion Exclusion
  • Wide Applicability (Versatility)
    • A wide range of samples can be analyzed with PRP-X300 columns because the selectivity of the column can be altered by changing the pH of the buffer or adding an organic modifier (Methanol, Acetonitrile) to the buffer. The supports stability to organic solvents makes it possible to analyze samples that have been too highly retained on conventional ion exclusion supports. The use of isocratic conditions allows samples to be analyzed without column reequilibration.
  • Fast Results
    • The excellent selectivity of the PRP-X300 columns ensures most separations are complete within five minutes
  • Easy Sample Preparation
    • Most samples require only minimal preparation before injection

    • One particle size: 7μm
    • Three column internal diameters: 1.0 to 4.6mm
    • Two column materials: 316 stainless steel and PEEK
    • Analytical and semiprep/preparative guard columns

    Specifications

    Column Format Analytical Column
    Diameter (Metric) 4.1 mm
    Length (Metric) 250 mm
    Max. Pressure 5000 psi (345 bar)
    Mobile Phase Compatibility pH 1 to 13, 0 to 100% aqueous, organic modifier
    Particle Size 7 μm
    pH 1 to 13
    Pore Size 100 Å
    Temperature 5°C to 60°C
    USP Type L17
    Stationary Phase PRP-X300
    Packing Material PSDVB/Sulfonic Acid
    Column Type Ion Exchange
    Phase Ion Exchange
    Material Stainless Steel
    Type Column
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.