Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSCA Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
$243.00 - $522.50
Specifications
Antigen | PSCA |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543623
|
Invitrogen™
MA543623 |
20 μL |
Each for $243.00
|
|
|||||
PIMA543622
|
Invitrogen™
MA543622 |
100 μL |
Each for $522.50
|
|
|||||
Description
PSCA (Prostate Stem Cell antigen) is a cell surface antigen, which is overexpressed in approximately40% of primary prostate cancers and in as many as 100% of metastatic ones. PSCA is also overexpressed in a majority of transitional cell and pancreatic carcinomas. Antibody directed against PSCA inhibits tumorigenesis, slows tumor growth, prolongs survival and prevents metastasis in a preclinical zenograft model indicating that PSCA may have utility as a prognostic marker and/or therapeutic target in prostate cancer.Specifications
PSCA | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
2210408B04Rik; caldesmon; CDM; HCAD; H-CAD; LCAD; L-CAD; NAG22; PRO232; prostate stem cell antigen; PSCA; UNQ206; UNQ206/PRO232 | |
PSCA | |
IgG1 κ | |
Protein A/G | |
Antibody |
C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O43653 | |
8000 | |
Leu13-Ser86 with N-terminal His Tag. | |
Primary | |
-20°C | |
PSCA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title