Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA4 Antibody (OTI4B8), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP273690V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273690V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273690V Supplier Novus Biologicals Supplier No. NBP273690V
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMA4 Monoclonal specifically detects PSMA4 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PSMA4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4B8
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HC9EC 3.4.25.1, HsT17706, Macropain subunit C9, MGC12467, MGC24813, Multicatalytic endopeptidase complex subunit C9, proteasome (prosome, macropain) subunit, alpha type, 4, Proteasome component C9, proteasome subunit alpha type-4, proteasome subunit HC9, Proteasome subunit L, PSC9MGC111191
Gene Symbols PSMA4
Host Species Mouse
Immunogen Full length recombinant protein of human PSMA4 (NP_002780) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 5685.0
Target Species Human, Mouse, Rat, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.