Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Pumice Specimen
SCI_ED

Catalog No. s26649 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Pumice is formed quickly during volcanic eruptions around pockets of air within lava.

  • Floats on water due to internal air pockets
  • For use in demonstrating effect of vesicles on rock density
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Glassy texture is result of swift cooling in mid-air

Specifications

Product Type Igneous Rock, Pumice
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.