Learn More
Eisco™ Pumice Specimen
SCI_ED
Catalog No. p-7076854 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
Pumice is formed quickly during volcanic eruptions around pockets of air within lava.
  • Floats on water due to internal air pockets
  • For use in demonstrating effect of vesicles on rock density
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Glassy texture is result of swift cooling in mid-air
Specifications
Product Type Igneous Rock, Pumice
For Use With (Application) Demonstrating effect of vesicles on rock density
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.