Learn More
Eisco™ Quartz Mineral Specimen
Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Description
- Useful during lessons on mineral hardness, luster, and fracture
- Has hardness of 7, complete lack of cleavage planes, and distinct euhedral crystal habit
Specifications
Specifications
| Product Type | Mineral, Quartz |
| For Use With (Application) | Study of mineral hardness, luster, and fracture |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 1 Ea. |
| Specimen Size | 2 to 3 cm |