Learn More
Eisco™ Quartz Mineral Specimen
SCI_ED
Catalog No. s24671 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Quartz mineral specimen for use in classroom identification labs and exercises.
  • Useful during lessons on mineral hardness, luster, and fracture
  • Has hardness of 7, complete lack of cleavage planes, and distinct euhedral crystal habit
Specifications
Product Type Mineral, Quartz
For Use With (Application) Study of mineral hardness, luster, and fracture
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.