Learn More
R&D Systems™ CellXVivo Human Th1 Cell Differentiation Kit
Catalog No. cdk001
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
The following optimized reagents to drive efficient differentiation of nanve CD4+ T cells into Th1 polarized cells: Mouse Anti-Human CD3 Antibody, Human Th1 Differentiation Supplements, Reconstitution Buffers, Wash Buffer (20X) Provides sufficient reagents for the differentiation of one 24-well plate or 250 tests in a 96-well format.
For the differentiation of Th1 cells from a preparation of naïve CD4+ T cells.
Specifications
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Quantity 1 Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.