Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB40B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $499.50

Specifications

Antigen RAB40B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15892220
SDP
View Documents
Novus Biologicals
NBP15892220UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP158922
SDP
View Documents
Novus Biologicals
NBP158922
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

RAB40B Polyclonal specifically detects RAB40B in Human samples. It is validated for Western Blot.
Specifications

Specifications

RAB40B
Polyclonal
Rabbit
Signal Transduction
GTP-binding protein homologous to Saccharomyces cerevisiae SEC4, Protein Rar, RAB40B, member RAS oncogene family, RAR, ras-related protein Rab-40B, SEC4LFLJ42385, SOCS box-containing protein RAR
RAB40B
IgG
Western Blot
Unconjugated
RUO
Q12829
10966
Synthetic peptides corresponding to RAB40B(RAB40B, member RAS oncogene family) The peptide sequence was selected from the middle region of RAB40B. Peptide sequence YAERLGVTFFEVSPLCNFNITESFTELARIVLLRHGMDRLWRPSKVLSLQ.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.