Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB6C Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RAB6C |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17426520
![]() |
Novus Biologicals
NBP17426520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174265
![]() |
Novus Biologicals
NBP174265 |
100 μL |
Each for $487.50
|
|
|||||
Description
RAB6C Polyclonal specifically detects RAB6C in Human samples. It is validated for Western Blot.Specifications
RAB6C | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
RAB6C, member RAS oncogene family, ras-related protein Rab-6C, WTH3Rab6-like protein WTH3 | |
RAB6C | |
IgG | |
28 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9H0N0 | |
84084 | |
Synthetic peptides corresponding to the middle region of RAB6C. Immunizing peptide sequence DVIITLVGNRTDLADKRQVSVEEGERKAKGLNVTFIETRAKAGYNVKQLF. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title